Proteintech
Manufacturer Code:157141AP150UL
Catalog # 50-173-0426
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NARS2 Fusion Protein Ag8345 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AsnRS; AsnRS Asparagine tRNA ligase NARS2 SLM5; asparagine tRNA ligase 2, mitochondrial (putative); asparagine--tRNA ligase; Asparaginyl-tRNA synthetase; asparaginyl-tRNA synthetase 2, mitochondrial; deafness, autosomal recessive 94; Nars2, similar to CG6796-PA; Probable asparagine--tRNA ligase, mitochondrial; probable asparaginyl-tRNA synthetase, mitochondrial
Gene Aliases: AI875199; asnRS; DFNB94; NARS2; RGD1305145; SLM5
UniProt ID: (Human) Q96I59, (Mouse) Q8BGV0
Entrez Gene ID: (Human) 79731, (Mouse) 244141, (Rat) 293128
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.