Invitrogen
Manufacturer Code:PA5116533
Catalog # PIPA5116533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 309-548 of human NARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3010001M15Rik AA960128 ASNRS asparagine tRNA ligase 1 cytoplasmic asparagine--tRNA ligase cytoplasmic Asparaginyl-tRNA synthetase asparaginyl-tRNA synthetase cytoplasmic C78150 LRRGT00113 NARS NARS1; AsnRS; asparagine tRNA ligase 1, cytoplasmic; Asparagine--tRNA ligase, cytoplasmic; Asparaginyl-tRNA synthetase; Asparaginyl-tRNA synthetase 1; asparaginyl-tRNA synthetase, cytoplasmic
Gene Aliases: 3010001M15Rik; AA960128; ASNRS; C78150; NARS; NARS1; NRS
UniProt ID: (Human) O43776, (Mouse) Q8BP47
Entrez Gene ID: (Human) 4677, (Mouse) 70223
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.