Novus Biologicals
Manufacturer Code:NBP219478
Catalog # NBP219478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human NARG1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ13340 GA19 Gastric cancer antigen Ga19 N(alpha)-acetyltransferase 15 NatA auxiliary subunit NARG1 NATHGa19 NMDA receptor-regulated protein 1 N-terminal acetyltransferase Protein tubedown-1 Tbdn100 TBDN100NatA auxiliary subunit transcriptional coactivator tubedown-100 tubedown-1; Gastric cancer antigen Ga19; N-alpha-acetyltransferase 15, NatA auxiliary subunit; N-terminal acetyltransferase; NMDA receptor regulated 1; NMDA receptor-regulated protein 1; Protein tubedown-1; Tbdn100; transcriptional coactivator tubedown-100; tubedown-1
Gene Aliases: GA19; NAA15; NARG1; NAT1P; NATH; TBDN; TBDN100
UniProt ID: (Human) Q9BXJ9
Entrez Gene ID: (Human) 80155
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.