Abnova
Manufacturer Code:PAB21591
Catalog # 89-123-088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human NARG1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant NARG1.
For Research Use Only
Protein Aliases: Ga19; Gastric cancer antigen Ga19; N-alpha-acetyltransferase 15, NatA auxiliary subunit; N-terminal acetyltransferase; NATH; NMDA receptor regulated 1; NMDA receptor-regulated protein 1; Protein tubedown-1; TBDN100; transcriptional coactivator tubedown-100; tubedown-1
Gene Aliases: GA19; NAA15; NARG1; NAT1P; NATH; TBDN; TBDN100
UniProt ID: (Human) Q9BXJ9
Entrez Gene ID: (Human) 80155
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.