OriGene
Manufacturer Code:CF808666
Catalog # 50-166-9006
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3E5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 64-244 of human NAPSA produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: asp 4; ASP4; Aspartyl protease 4; KAP Kdap NAP1 NAPA SNAPA; kidney-derived aspartic protease-like protein; Napsin-1; Napsin-A; pronapsin A; TA01/TA02
Gene Aliases: KAP; Kdap; NAP1; NAPA; NAPSA; SNAPA
UniProt ID: (Human) O96009
Entrez Gene ID: (Human) 9476
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.