Invitrogen
Manufacturer Code:PA570595
Catalog # PIPA570595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human NAPRT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FHA-HIT-interacting protein; NAPRTase; Nicotinate phosphoribosyltransferase; nicotinate phosphoribosyltransferase domain containing 1; Nicotinate phosphoribosyltransferase domain-containing protein 1; nicotinate phosphoribosyltransferase-like protein; nicotinic acid phosphoribosyltransferase; PP3856 FHA-HIT-interacting protein NAPRTase nicotinate phosphoribosyltransferase nicotinate phosphoribosyltransferase domain-containing protein 1 nicotinic acid phosphoribosyltransferase
Gene Aliases: 9130210N20Rik; FHIP; NAPRT; NAPRT1; PP3856; zgc:77870
UniProt ID: (Bovine) A5PK51, (Human) Q6XQN6, (Mouse) Q8CC86, (Rat) Q6XQN1
Entrez Gene ID: (Dog) 609412, (Pig) 403161, (Bovine) 399559, (Human) 93100, (Horse) 100147366, (Mouse) 223646, (Guinea pig) 100730251, (Rat) 315085, (Zebrafish) 393864
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.