Invitrogen
Manufacturer Code:PA5110012
Catalog # PIPA5110012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-312 of human NAPG (NP_003817.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2400003O04Rik GAMMASNAP Gamma-soluble NSF attachment protein Napg N-ethylmaleimide-sensitive factor attachment protein gamma SNAPG SNAP-gamma SNARE; gamma SNAP; Gamma-soluble NSF attachment protein; N-ethylmaleimide sensitive fusion protein attachment protein gamma; N-ethylmaleimide-sensitive factor attachment protein gamma; N-ethylmaleimide-sensitive factor attachment protein, gamma; SNAP-gamma; soluble NSF attachment protein
Gene Aliases: 2400003O04Rik; GAMMASNAP; NAPG; SNAPG; SNARE
UniProt ID: (Human) Q99747, (Mouse) Q9CWZ7
Entrez Gene ID: (Human) 8774, (Rat) 307382, (Mouse) 108123
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.