Novus Biologicals
Manufacturer Code:NB10075437
Catalog # NB10075437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody was a sequence from 1a.a. to 268a.a. from NAPG protein(a 312a.a. long protein) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: gamma SNAP; gamma SNAP GAMMASNAP gamma-soluble NSF attachment protein N-ethylmaleimide-sensitive factor attachment protein gamma N-ethylmaleimide-sensitive factor attachment protein gamma SNAPG SNAP-gamma soluble NSF attachment protein; Gamma-soluble NSF attachment protein; N-ethylmaleimide-sensitive factor attachment protein gamma; N-ethylmaleimide-sensitive factor attachment protein, gamma; SNAP-gamma; soluble NSF attachment protein
Gene Aliases: GAMMASNAP; NAPG; SNAPG
UniProt ID: (Human) Q99747
Entrez Gene ID: (Human) 8774
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.