Invitrogen
Manufacturer Code:PA5115616
Catalog # PIPA5115616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NAPE-PLD corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A530089G06 C7orf18 DKFZp781D1098 FMP30 Mbldc1 Metallo-beta-lactamase domain containing 1 N-acyl phosphatidylethanolamine phospholipase D N-acyl-phosphatidylethanolamine-hydrolyzing phospholipase D NAPE-hydrolyzing phospholipase D NAPEPLD NAPE-PLD; Metallo-beta-lactamase domain containing 1; N-acyl phosphatidylethanolamine phospholipase D; N-acyl-phosphatidylethanolamine-hydrolyzing phospholipase D; NAPE-hydrolyzing phospholipase D
Gene Aliases: A530089G06; C7orf18; FMP30; Mbldc1; NAPE-PLD; NAPEPLD
UniProt ID: (Human) Q6IQ20, (Mouse) Q8BH82, (Rat) Q769K2
Entrez Gene ID: (Human) 222236, (Mouse) 242864, (Rat) 296757
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.