Novus Biologicals
Manufacturer Code:NB11080070
Catalog # NB11080070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of mouse NAPE-PLD (within residues 1-100). [Swiss-Prot# Q8BH82] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C7orf18 DKFZp781D1098 EC 3.1.4 EC 3.1.4.- N-acyl phosphatidylethanolamine phospholipase DNAPE-PLDFMP30 N-acyl-phosphatidylethanolamine-hydrolyzing phospholipase D NAPE-hydrolyzing phospholipase D; N-acyl phosphatidylethanolamine phospholipase D; N-acyl-phosphatidylethanolamine-hydrolyzing phospholipase D; NAPE-hydrolyzing phospholipase D
Gene Aliases: C7orf18; FMP30; NAPE-PLD; NAPEPLD
UniProt ID: (Human) Q6IQ20
Entrez Gene ID: (Human) 222236
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.