Invitrogen
Manufacturer Code:PA5116807
Catalog # PIPA5116807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human NAPB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta soluble NSF attachment protein; beta soluble NSF attachment protein beta-soluble NSF attachment protein beta-soluble NSF attachment protein {ECO:0000250 brain protein 14 brain protein I47 Brp14 b-SNAP E161 I47 Napb napb {ECO:0000250 N-ethylmaleimide sensitive fusion protein attachment protein beta N-ethylmaleimide-sensitive factor attachment protein beta N-ethylmaleimide-sensitive factor attachment protein beta {ECO:0000250 N-ethylmaleimide-sensitive factor attachment protein beta NSF attachment protein beta Snapb SNAP-beta SNAP-beta {ECO:0000250 SNARE UniProtKB:P28663}; Beta-soluble NSF attachment protein; beta-soluble NSF attachment protein {ECO:0000250|UniProtKB:P28663}; brain protein 14; Brain protein I47; N-ethylmaleimide sensitive fusion protein attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta {ECO:0000250|UniProtKB:P28663}; N-ethylmaleimide-sensitive factor attachment protein, beta; napb {ECO:0000250|UniProtKB:P28663}; SNAP-beta; SNAP-beta {ECO:0000250|UniProtKB:P28663}
Gene Aliases: b-SNAP; Brp14; E161; I47; NAPB; SNAP-BETA; SNAPB; SNARE
UniProt ID: (Human) Q9H115, (Mouse) P28663
Entrez Gene ID: (Human) 63908, (Rat) 499903, (Mouse) 17957
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.