Novus Biologicals
Manufacturer Code:NBP294485
Catalog # NB023503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human NAPB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta soluble NSF attachment protein; Beta-soluble NSF attachment protein; MGC26066 N-ethylmaleimide-sensitive factor attachment protein beta SNAP-BETA; N-ethylmaleimide-sensitive factor attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein, beta; SNAP-beta
Gene Aliases: NAPB; SNAP-BETA; SNAPB
UniProt ID: (Human) Q9H115
Entrez Gene ID: (Human) 63908
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.