Invitrogen
Manufacturer Code:PA569910
Catalog # PIPA569910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human NANP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf147 HDHD4 dJ694B14.3 N-acylneuraminate-9-phosphatase Neu5Ac-9-Pase haloacid dehalogenase-like hydrolase domain containing 4 haloacid dehalogenase-like hydrolase domain-containing protein 4; haloacid dehalogenase-like hydrolase domain containing 4; Haloacid dehalogenase-like hydrolase domain-containing protein 4; N-acylneuraminate-9-phosphatase; nanp {ECO:0000250|UniProtKB:Q9CPT3}; Neu5Ac-9-Pase
Gene Aliases: 1600031M04Rik; C20orf147; dJ694B14.3; HDHD4; NANP; RGD1306009; zgc:111947
UniProt ID: (Human) Q8TBE9, (Rat) Q5M969, (Mouse) Q9CPT3
Entrez Gene ID: (Bovine) 516539, (Pig) 100518994, (Rabbit) 100341646, (Dog) 608182, (Human) 140838, (Rat) 311530, (Guinea pig) 100720176, (Mouse) 67311, (Zebrafish) 553795
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.