Abnova
Manufacturer Code:PAB16304
Catalog # 89-118-178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human NANP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NANP.
For Research Use Only
Protein Aliases: C20orf147; dJ694B14.3; haloacid dehalogenase-like hydrolase domain containing 4; Haloacid dehalogenase-like hydrolase domain-containing protein 4; HDHD4; MGC26833; N-acylneuraminate-9-phosphatase; Neu5Ac-9-Pase
Gene Aliases: C20orf147; dJ694B14.3; HDHD4; NANP
UniProt ID: (Human) Q8TBE9
Entrez Gene ID: (Human) 140838
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.