Novus Biologicals
Manufacturer Code:NBP177327
Catalog # NBP177327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human Nanos1. The immunogen is located within the first 50 amino acids of Nanos1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: EC_Rep1a; Nanos homolog 1; nanos homolog 1 nanos homolog 1 (Drosophila) NOS-1 NOS1EC_Rep1a; NOS-1
Gene Aliases: EC_Rep1a; NANOS1; NOS-1; NOS1; SPGF12; ZC2HC12A
UniProt ID: (Human) Q8WY41
Entrez Gene ID: (Human) 340719
Molecular Function:
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.