Novus Biologicals
Manufacturer Code:NBP232538
Catalog # NBP232538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: TILANKKQSAMPWDQDPEQSTGNYSEDEQNGKQKWREEGEAGRKRE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeobox C14; Homeobox C14 Homeobox Protein C14 NANOG Neighbor Homeobox; Homeobox protein C14; NANOG neighbor homeobox
Gene Aliases: NANOGNB
UniProt ID: (Human) Q7Z5D8
Entrez Gene ID: (Human) 360030
Molecular Function:
cytoskeletal protein
hydrolase
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.