Abnova
Manufacturer Code:PAB13693
Catalog # 89-116-807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse Nanog protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against Nanog.
For Research Use Only
Protein Aliases: 2410002E02Rik; early embryo specific expression NK family; Early embryo specific expression NK-type homeobox protein; ecat4; ENK; ES cell-associated protein 4; Homeobox protein NANOG; Homeobox transcription factor Nanog
Gene Aliases: 2410002E02Rik; Ecat4; ENK; Nanog
UniProt ID: (Mouse) Q80Z64
Entrez Gene ID: (Mouse) 71950
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.