Abnova
Manufacturer Code:MAB1122
Catalog # 89-109-390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D5 |
Immunogen | Recombinant protein corresponding to full length human NAMPT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full-length recombinant NAMPT.
For Research Use Only
Protein Aliases: 1110035O14Rik; DKFZp666B131; MGC117256; NAmPRTase; Nicotinamide phosphoribosyltransferase; PBEF; PBEF1; Pre-B cell-enhancing factor; pre-B-cell colony enhancing factor 1; Pre-B-cell colony-enhancing factor 1; VF; VISFATIN
Gene Aliases: 1110035O14Rik; NAMPT; PBEF; PBEF1; VF; VISFATIN
UniProt ID: (Human) P43490
Entrez Gene ID: (Human) 10135
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.