Invitrogen
Manufacturer Code:710115
Catalog # PI710115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 21HCLC |
Immunogen | Recombit protein corresponding to amino acids 1110 of human Visfatin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: NAmPRTase; Nicotinamide phosphoribosyltransferase; PBEF PBEF1 VF NAmPRTase pre-B cell-enhancing factor pre-B-cell colony enhancing factor 1 pre-B-cell colony-enhancing factor 1; Pre-B cell-enhancing factor; pre-B-cell colony enhancing factor 1; Pre-B-cell colony-enhancing factor 1; Visfatin
Gene Aliases: 1110035O14Rik; NAMPT; PBEF; PBEF1; VF; VISFATIN
UniProt ID: (Human) P43490
Entrez Gene ID: (Human) 10135
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.