Abnova
Manufacturer Code:PAB9553
Catalog # 89-134-226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to mouse Nampt. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a recombinant Nampt.
For Research Use Only
Protein Aliases: 1110035O14Rik; AI314458; AI480535; NAmPRTase; Nicotinamide phosphoribosyltransferase; PBEF; Pbef1; Pre-B-cell colony-enhancing factor 1 homolog; Visfatin
Gene Aliases: 1110035O14Rik; AI314458; AI480535; NAmPRTase; Nampt; Pbef; Pbef1; Visfatin
UniProt ID: (Mouse) Q99KQ4
Entrez Gene ID: (Mouse) 59027
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.