Novus Biologicals
Manufacturer Code:NBP231372SS
Catalog # NBP231372SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Partial recombinant protein made to an N-terminal portion of human NALP6 (between amino acids 10-200) [UniProt P59044]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Angiotensin II/vasopressin receptor; Angiotensin II/Vasopressin Receptor AVR CLR11.4 NACHT Leucine Rich Repeat And PYD Containing 6 NACHT LRR And PYD Containing Protein 6 NACHT LRR And PYD Domains-Containing Protein 6 NAVR NAVR/AVR NLR Family Pyrin Domain Containing 6 NLRP6 Nucleotide-Binding Oligomerization Domain Leucine Rich Repeat And Pyrin Domain Containing 6 PAN3 PYPAF5 PYRIN-Containing APAF1-Like Protein 5; NACHT, leucine rich repeat and PYD containing 6; NACHT, LRR and PYD domains-containing protein 6; NLR family, pyrin domain containing 6; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 6; PYRIN-containing APAF1-like protein 5
Gene Aliases: AVR; CLR11.4; NALP6; NAVR; NAVR/AVR; NLRP6; PAN3; PYPAF5
UniProt ID: (Human) P59044
Entrez Gene ID: (Human) 171389
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.