Invitrogen
Manufacturer Code:PA572910
Catalog # PIPA572910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 139-157 of human NALP4/PAN2/PYPAF4 was used as immunogen GenBank no. gi 17064172 gb AAL35293.1 AF442488&lowbar1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 58; CLR19.5 CT58 NALP4 PAN2 PYPAF4 RNH2 NACHT LRR and PYD containing protein 4 NACHT LRR and PYD domains-containing protein 4 NACHT leucine rich repeat and PYD containing 4 PAAD and NACHT-containing protein 2 PYRIN and NACHT-containing protein 2 PYRIN-containing APAF1-like protein 4 cancer/testis antigen 58 nucleotide-binding oligomerization domain leucine rich repeat and pyrin domain containing 4 ribonuclease inhibitor 2; CT58; NACHT, leucine rich repeat and PYD containing 4; NACHT, leucine rich repeat and PYD containing 4E; NACHT, LRR and PYD containing protein 4; NACHT, LRR and PYD domains-containing protein 4; NACHT, LRR and PYD domains-containing protein 4E; NALP, epsilon; NALP-epsilon; NLR family, pyrin domain containing 4; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 4; PAAD and NACHT-containing protein 2; PAN2; PYPAF4; PYRIN and NACHT-containing protein 2; PYRIN-containing APAF1-like protein 4; Ribonuclease inhibitor 2
Gene Aliases: 4930406H16Rik; CLR19.5; CT58; Nalp-epsilon; NALP4; Nalp4e; NLRP4; Nlrp4e; PAN2; PYPAF4; RNH2
UniProt ID: (Human) Q96MN2, (Mouse) Q66X19
Entrez Gene ID: (Human) 147945, (Mouse) 446099
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.