Novus Biologicals
Manufacturer Code:NB10056156SS
Catalog # NB10056156T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 139-157 (FAPKETGKQPRTVIIQGPQ) of human NALP4/PAN2/PYPAF4 was used as immunogen GenBank no. gi 17064172 gb AAL35293.1 AF442488_1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 58; Cancer/testis antigen 58 CLR19.5 CT58PYRIN and NACHT-containing protein 2 FLJ32126 NACHT leucine rich repeat and PYD containing 4 NACHT LRR and PYD domains-containing protein 4 NALP4NACHT LRR and PYD containing protein 4 NLR family pyrin domain containing 4 nucleotide-binding oligomerization domain leucine rich repeat and pyrin domaincontaining 4 PAN2Ribonuclease inhibitor 2 PYPAF4PAAD and NACHT-containing protein 2 RNH2PYRIN-containing APAF1-like protein 4; CT58; NACHT, leucine rich repeat and PYD containing 4; NACHT, LRR and PYD containing protein 4; NACHT, LRR and PYD domains-containing protein 4; NLR family, pyrin domain containing 4; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 4; PAAD and NACHT-containing protein 2; PAN2; PYPAF4; PYRIN and NACHT-containing protein 2; PYRIN-containing APAF1-like protein 4; Ribonuclease inhibitor 2
Gene Aliases: CLR19.5; CT58; NALP4; NLRP4; PAN2; PYPAF4; RNH2
UniProt ID: (Human) Q96MN2
Entrez Gene ID: (Human) 147945
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.