Invitrogen
Manufacturer Code:PA523311
Catalog # PIPA523311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (493 REVQRLLEVKEQEGNDE 509) of NALP10 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLR11.1 NALP10 NOD8 PAN5 PYNOD NACHT LRR and PYD domains-containing protein 10 NACHT leucine rich repeat and PYD containing 10 nucleotide-binding oligomerization domain protein 8 nucleotide-binding oligomerization domain leucine rich repeat and pyrin domain containing 10; NACHT, leucine rich repeat and PYD containing 10; NACHT, LRR and PYD containing protein 10; NACHT, LRR and PYD domains-containing protein 10; NACHT/LRR/pyrin domain-containing protein 10; neuronal apoptosis inhibitor protein 10; NLR family, pyrin domain containing 10; Nucleotide-binding oligomerization domain protein 8; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 10
Gene Aliases: 6430548I20Rik; CLR11.1; NALP10; Napl10; NLRP10; NOD8; PAN5; PYNOD
UniProt ID: (Human) Q86W26, (Mouse) Q8CCN1
Entrez Gene ID: (Human) 338322, (Mouse) 244202
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.