Invitrogen
Manufacturer Code:PA5113073
Catalog # PIPA5113073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human NACHT LRR and PYD domains-containing protein 10 protein (589-655AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6430548I20Rik CLR11.1 NACHT leucine rich repeat and PYD containing 10 NACHT LRR and PYD containing protein 10 NACHT LRR and PYD domains-containing protein 10 NACHT/LRR/pyrin domain-containing protein 10 Nalp10 Napl10 neuronal apoptosis inhibitor protein 10 NLR family pyrin domain containing 10 NLR family pyrin domain containing 10 NLRP10 NOD8 Nucleotide-binding oligomerization domain protein 8 nucleotide-binding oligomerization domain leucine rich repeat and pyrin domain containing 10 PAN5 Pynod; NACHT, leucine rich repeat and PYD containing 10; NACHT, LRR and PYD domains-containing protein 10; NLR family, pyrin domain containing 10; Nucleotide-binding oligomerization domain protein 8; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 10
Gene Aliases: CLR11.1; NALP10; NLRP10; NOD8; PAN5; PYNOD
UniProt ID: (Human) Q86W26
Entrez Gene ID: (Human) 338322, (Rat) 293426
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.