Invitrogen
Manufacturer Code:PA5116672
Catalog # PIPA5116672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1000-1100 of human NLRP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Card7 caspase recruitment domain protein caspase recruitment domain protein 7 caspase recruitment domain-containing protein 7 CIDED CLR17.1 Death effector filament-forming ced-4-like apoptosis protein DEFCAP DEFCAP-L/S DKFZp586O1822 Gm14 Gm15 KIAA0926 NAC nacht NACHT leucine rich repeat and PYD (pyrin domain) containing 1 NACHT leucine rich repeat and PYD containing 1 NACHT LRR and PYD containing protein 1 NACHT LRR and PYD domains-containing protein 1 NACHT LRR and PYD domains-containing protein 1a NACHT/LRR/pyrin domain-containing protein 1 Nalp1 Nalp1a NLR family protein 1 NLR family pyrin domain containing 1 NLR family pyrin domain containing 1 NLR family pyrin domain containing 1A NLRP1 Nlrp1a nucleotide-binding domain and caspase recruitment domain nucleotide-binding oligomerization domain leucine rich repeat and pyrin domain containing 1 PP1044 resistant anthrax lethal toxin SLEV1 VAMAS1; caspase recruitment domain protein; caspase recruitment domain protein 7; Caspase recruitment domain-containing protein 7; Death effector filament-forming ced-4-like apoptosis protein; NACHT, leucine rich repeat and PYD (pyrin domain) containing 1; NACHT, leucine rich repeat and PYD containing 1; NACHT, LRR and PYD containing protein 1; NACHT, LRR and PYD domains-containing protein 1; NACHT, LRR and PYD domains-containing protein 1a; NACHT/LRR/pyrin domain-containing protein 1; NLR family protein 1; NLR family, pyrin domain containing 1; NLR family, pyrin domain containing 1A; Nucleotide-binding domain and caspase recruitment domain; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 1; resistant anthrax lethal toxin
Gene Aliases: CARD7; CIDED; CLR17.1; DEFCAP; DEFCAP-L/S; Gm14; Gm15; KIAA0926; NAC; NALP1; Nalp1a; NLRP1; Nlrp1a; PP1044; SLEV1; VAMAS1
UniProt ID: (Human) Q9C000, (Mouse) Q2LKU9
Entrez Gene ID: (Human) 22861, (Mouse) 195046, (Rat) 360557
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.