Invitrogen
Manufacturer Code:MA527593
Catalog # PIMA527593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S187-7 |
Immunogen | Fusion protein amino acids 1659-1738 (cytoplasmic C-terminus) of rat NALCN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: brain voltage-gated cation channel; CanIon; Four domain-type voltage-gated ion channel alpha-1 subunit; four repeat voltage-gated ion channel; Rb21-channel; Sodium leak channel non-selective protein; sodium leak channel, non selective; VGCNL1 CanIon Sodium leak channel non-selective protein Voltage gated channel-like protein 1 A530023G15Rik bA430M15.1 FLJ23913 FLJ44659 FLJ44764 Four repeat voltage gated ion channel MGC74524 Putative 4 repeat voltage gated ion channel Sodium leak channel non-selective protein; voltage gated channel like 1; Voltage gated channel-like protein 1
Gene Aliases: A530023G15Rik; AI849508; bA430M15.1; CanIon; CLIFAHDD; IHPRF; IHPRF1; INNFD; NALCN; Nca; Rb21; VGCNL1
UniProt ID: (Human) Q8IZF0, (Mouse) Q8BXR5, (Rat) Q6Q760
Entrez Gene ID: (Human) 259232, (Mouse) 338370, (Rat) 266760
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.