Abnova
Manufacturer Code:H00259232K
Catalog # 89-206-609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human NALCN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human NALCN peptide using ARM Technology.
For Research Use Only
Protein Aliases: CanIon; CanIon FLJ23913 FLJ44659 FLJ44764 MGC74524 VGCNL1 bA430M15.1; four repeat voltage-gated ion channel; Sodium leak channel non-selective protein; sodium leak channel, non selective; voltage gated channel like 1; Voltage gated channel-like protein 1
Gene Aliases: bA430M15.1; CanIon; CLIFAHDD; IHPRF; IHPRF1; INNFD; NALCN; VGCNL1
UniProt ID: (Human) Q8IZF0
Entrez Gene ID: (Human) 259232
Molecular Function:
calcium channel
ion channel
sodium channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.