R&D Systems
Manufacturer Code:MAB829
Catalog # MAB829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 541609 |
Immunogen | <em>E.coli</em>-derived recombinant human NAIP Asn923-Val1148 Accession # Q13075 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: baculoviral IAP repeat-containing 1 BIRC1 BIRC1baculoviral IAP repeat-containing protein 1 FLJ18088 FLJ42520 FLJ58811 Neuronal apoptosis inhibitory protein NLR family apoptosis inhibitory protein NLR family BIR domain containing 1 NLRB1 nucleotide-binding oligomerization domain leucine rich repeat and BIR domaincontaining 1 psi neuronal apoptosis inhibitory protein psiNAIP; Baculoviral IAP repeat-containing protein 1; Neuronal apoptosis inhibitory protein; NLR family, apoptosis inhibitory protein; nucleotide-binding oligomerization domain, leucine rich repeat and BIR domain containing 1; psi neuronal apoptosis inhibitory protein
Gene Aliases: BIRC1; NAIP; NLRB1; psiNAIP
UniProt ID: (Human) Q13075
Entrez Gene ID: (Human) 4671
Molecular Function:
enzyme modulator
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.