Invitrogen
Manufacturer Code:PA559978
Catalog # PIPA559978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NAIP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Baculoviral IAP repeat-containing protein 1; BIRC1 NLRB1 baculoviral IAP repeat-containing 1 BIRC1baculoviral IAP repeat-containing protein 1 FLJ18088 FLJ42520 FLJ58811 Neuronal apoptosis inhibitory protein NLR family apoptosis inhibitory protein NLR family BIR domain containing 1 NLRB1 nucleotide-binding oligomerization domain leucine rich repeat and BIR domaincontaining 1 psi neuronal apoptosis inhibitory protein psiNAIP; Neuronal apoptosis inhibitory protein; NLR family, apoptosis inhibitory protein; nucleotide-binding oligomerization domain, leucine rich repeat and BIR domain containing 1; psi neuronal apoptosis inhibitory protein
Gene Aliases: BIRC1; NAIP; NLRB1; psiNAIP
UniProt ID: (Human) Q13075
Entrez Gene ID: (Human) 4671
Molecular Function:
enzyme modulator
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.