Invitrogen
Manufacturer Code:PA5115614
Catalog # PIPA5115614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NAIP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AV364616 baculoviral IAP repeat-containing 1a baculoviral IAP repeat-containing protein 1 Baculoviral IAP repeat-containing protein 1a BIRC1 Birc1a D13Lsd1 FLJ18088 FLJ42520 FLJ58811 Naip Naip1 Naip-rs1 neuronal apoptosis inhibitory protein Neuronal apoptosis inhibitory protein 1 NLR family apoptosis inhibitory protein NLR family apoptosis inhibitory protein NLR family apoptosis inhibitory protein 1 NLRB1 nucleotide-binding oligomerization domain leucine rich repeat and BIR domain containing 1 psi neuronal apoptosis inhibitory protein psiNAIP; baculoviral IAP repeat-containing 1a; Baculoviral IAP repeat-containing protein 1; Baculoviral IAP repeat-containing protein 1a; Neuronal apoptosis inhibitory protein; Neuronal apoptosis inhibitory protein 1; NLR family, apoptosis inhibitory protein; nucleotide-binding oligomerization domain, leucine rich repeat and BIR domain containing 1; psi neuronal apoptosis inhibitory protein
Gene Aliases: AV364616; BIRC1; Birc1a; D13Lsd1; NAIP; Naip-rs1; Naip1; NLRB1; psiNAIP
UniProt ID: (Human) Q13075, (Mouse) Q9QWK5
Entrez Gene ID: (Human) 4671, (Mouse) 17940
Molecular Function:
enzyme modulator
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.