Novus Biologicals
Manufacturer Code:NBP179444
Catalog # NBP179444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal of human NAGSThe immunogen for this antibody is NAGS. Peptide sequence YLDKFVVSSSRQGQGSGQMLWECLRRDLQTLFWRSRVTNPINPWYFKHSD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGAS Amino-acid acetyltransferase ARGA EC 2.3.1.1 MGC133025 N-acetylglutamate synthase N-acetylglutamate synthase mitochondrial NAT7; Amino-acid acetyltransferase; N-acetylglutamate synthase conserved domain form; N-acetylglutamate synthase long form; N-acetylglutamate synthase short form; N-acetylglutamate synthase, mitochondrial
Gene Aliases: AGAS; ARGA; NAGS
UniProt ID: (Human) Q8N159
Entrez Gene ID: (Human) 162417
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.