Abnova
Manufacturer Code:PAB4634
Catalog # 89-131-503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human NAGK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NAGK.
For Research Use Only
Protein Aliases: GlcNAc kinase; GNK; HSA242910; N-acetyl-D-glucosamine kinase; N-acetylglucosamine kinase
Gene Aliases: GNK; HSA242910; NAGK
UniProt ID: (Human) Q9UJ70
Entrez Gene ID: (Human) 55577
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.