Proteintech
Manufacturer Code:150511AP150UL
Catalog # 50-172-9933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NAGK Fusion Protein Ag7034 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GlcNAc kinase; GlcNAc kinase GNK HSA242910 N acetyl D glucosamine kinase N acetylglucosamine kinase NAGK; N-acetyl-D-glucosamine kinase; N-acetylglucosamine kinase
Gene Aliases: GNK; HSA242910; NAGK
UniProt ID: (Human) Q9UJ70, (Mouse) Q9QZ08, (Rat) P81799
Entrez Gene ID: (Human) 55577, (Mouse) 56174, (Rat) 297393
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.