Invitrogen
Manufacturer Code:PA576459
Catalog # PIPA576459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human NAGA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylgalactosaminidase, alpha-N- (alpha-galactosidase B); Alpha-galactosidase B; Alpha-galactosidase B alpha-galactosidase B galactosidasealphaB galactosidase-alphaB; Alpha-N-acetylgalactosaminidase; N-acetylgalactosaminidase, alpha-
Gene Aliases: D22S674; GALB; NAGA
UniProt ID: (Human) P17050, (Rat) Q66H12, (Mouse) Q9QWR8
Entrez Gene ID: (Human) 4668, (Rat) 315165, (Mouse) 17939
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.