Invitrogen
Manufacturer Code:PA527690
Catalog # PIPA527690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 111 of Human NADE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BEX3 Bex DXS6984E HGR74 NADE brain expressed X-linked 3 brain-expressed X-linked protein 3 nerve growth factor receptor-associated protein 1 ovarian granulosa cell 13.0 kDa protein HGR74 ovarian granulosa cell protein (13kD) p75NTR-associated cell death executor protein BEX3; Brain-expressed X-linked protein 3; nerve growth factor receptor (TNFRSF16) associated protein 1; Nerve growth factor receptor-associated protein 1; Ovarian granulosa cell 13.0 kDa protein HGR74; p75NTR-associated cell death executor; Protein BEX3
Gene Aliases: Bex; BEX3; DXS6984E; HGR74; NADE; NGFRAP1
UniProt ID: (Human) Q00994
Entrez Gene ID: (Human) 27018
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.