Invitrogen
Manufacturer Code:PA520076
Catalog # PIPA520076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide corresponding to 14 amino acids near the middle of human NADE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain expressed X-linked 3; Brain-expressed X-linked protein 3; Brain-expressed X-linked protein 3 homolog; granule cell antiserum positive 27; nerve growth factor receptor (TNFRSF16) associated protein 1; Nerve growth factor receptor-associated protein 1; Ovarian granulosa cell 13.0 kDa protein HGR74; p75NTR-associated Cell Death Executor; Protein BEX3; RP13-349O20.1 BEX3 Bex DXS6984E HGR74 NADE brain expressed X-linked 3 brain-expressed X-linked protein 3 nerve growth factor receptor-associated protein 1 ovarian granulosa cell 13.0 kDa protein HGR74 ovarian granulosa cell protein (13kD) p75NTR-associated cell death executor protein BEX3
Gene Aliases: AL033356; Bex; BEX3; DXS6984E; DXWsu67e; Gcap27; HGR74; NADE; NGFRAP1
UniProt ID: (Human) Q00994, (Mouse) Q9WTZ9
Entrez Gene ID: (Human) 27018, (Mouse) 12070
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.