Invitrogen
Manufacturer Code:PA572429
Catalog # PIPA572429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | region 351 - 410 aa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0363; NAC-alpha domain-containing protein 1
Gene Aliases: D230024G13Rik; KIAA0363; mKIAA0363; NACAD
UniProt ID: (Human) O15069, (Mouse) Q5SWP3
Entrez Gene ID: (Human) 23148, (Rat) 289786, (Mouse) 192950
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.