Novus Biologicals
Manufacturer Code:NBP29338920UL
Catalog # NB028661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 278-487 of human NAB1 (NP_005957.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EGR-1-binding protein 1; EGR1 binding protein 1; EGR1 binding protein 1 EGR-1-binding protein 1 NGFI-A binding protein 1 (EGR1 binding protein 1) NGFI-A-binding protein 1 Transcriptional regulatory protein p54; NGFI-A binding protein 1 (EGR1 binding protein 1); NGFI-A-binding protein 1; Transcriptional regulatory protein p54
Gene Aliases: NAB1
UniProt ID: (Human) Q13506
Entrez Gene ID: (Human) 4664
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.