Invitrogen
Manufacturer Code:PA570792
Catalog # PIPA570792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of mouse NAALADL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutamate carboxypeptidase II-type non-peptidase homologue; Inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2; N-acetylated alpha-linked acidic dipeptidase 2; N-acetylated alpha-linked acidic dipeptidase-like 2; NAALADase L2; NAALADL2 inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2 NAALADase L2 N-acetylated alpha-linked acidic dipeptidase 2 N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases: NAALADL2; RGD1565147
UniProt ID: (Human) Q58DX5
Entrez Gene ID: (Dog) 488174, (Horse) 100064889, (Human) 254827, (Rabbit) 100342215, (Rat) 499583, (Guinea pig) 100719214
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.