Invitrogen
Manufacturer Code:PA5112966
Catalog # PIPA5112966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2 protein (143-795AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810043G22Rik EG635702 glutamate carboxypeptidase II-type non-peptidase homologue Gm1021 Inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2 NAALADase L2 NAALADL2 N-acetylated alpha-linked acidic dipeptidase 2 N-acetylated alpha-linked acidic dipeptidase like 2 N-acetylated alpha-linked acidic dipeptidase-like 2 RGD1565147; glutamate carboxypeptidase II-type non-peptidase homologue; Inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2; N-acetylated alpha-linked acidic dipeptidase 2; NAALADase L2
Gene Aliases: NAALADL2
UniProt ID: (Human) Q58DX5
Entrez Gene ID: (Human) 254827
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.