Invitrogen
Manufacturer Code:MA529434
Catalog # PIMA529434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 29 |
Immunogen | Recombit Human NAALADL1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa ileum brush border membrane protein; Aminopeptidase NAALADL1; I100; Ileal dipeptidylpeptidase; ileal dipeptidylpeptidase NAALADase L NAALADase-like 1 NAALADL1 I100 NAALADASEL NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1; ileal peptidase I100; N-acetylated-alpha-linked acidic dipeptidase-like protein; NAALADase L
Gene Aliases: I100; NAALADASEL; NAALADL; NAALADL1
UniProt ID: (Human) Q9UQQ1
Entrez Gene ID: (Human) 10004
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.