Invitrogen
Manufacturer Code:PA5107003
Catalog # PIPA5107003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: EQVRLLSSSL TADCSLRSPS GREVEPGEDR TIRYVRYESE LQMPDIMRLI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: N-acetyltransferase 12; N-acetyltransferase 12 (GCN5-related, putative); N-acetyltransferase MAK3 homolog; N-alpha-acetyltransferase 30; N-alpha-acetyltransferase 30, NatC catalytic subunit; NatC catalytic subunit; putative N-acetyltransferase
Gene Aliases: C14orf35; MAK3; Mak3p; NAA30; NAT12; NAT12P
UniProt ID: (Human) Q147X3
Entrez Gene ID: (Human) 122830
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.