Invitrogen
Manufacturer Code:PA543248
Catalog # PIPA543248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human NARG1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: N(alpha)-acetyltransferase 16, NatA auxiliary subunit; N-alpha-acetyltransferase 16, NatA auxiliary subunit; Nalpha acetyltransferase 16; NARG1-like protein; NMDA receptor regulated 1-like protein; NMDA receptor-regulated 1-like protein; NMDA receptor-regulated 1-like protein-like protein
Gene Aliases: 1300019C06Rik; NAA16; NARG1L; NAT2
UniProt ID: (Human) Q6N069, (Mouse) Q9DBB4
Entrez Gene ID: (Horse) 100061122, (Human) 79612, (Bovine) 538989, (Pig) 100158125, (Dog) 485464, (Rabbit) 100342198, (Mouse) 66897, (Rat) 686240
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.