Invitrogen
Manufacturer Code:PA531688
Catalog # PIPA531688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 187 of Human NARG1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: N-alpha-acetyltransferase 16, NatA auxiliary subunit; NARG1-like protein; NARG1L N-alpha-acetyltransferase 16 NatA auxiliary subunit NARG1-like protein NMDA receptor-regulated 1-like protein; NMDA receptor-regulated 1-like protein
Gene Aliases: NAA16; NARG1L; NAT2
UniProt ID: (Human) Q6N069
Entrez Gene ID: (Human) 79612
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.