Invitrogen
Manufacturer Code:PA530424
Catalog # PIPA530424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 312 of Human NARG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ga19 NARG1 NATH TBDN100 N-alpha-acetyltransferase 15 NatA auxiliary subunit N-terminal acetyltransferase NMDA receptor regulated 1 NMDA receptor-regulated protein 1 gastric cancer antigen Ga19 protein tubedown-1 transcriptional coactivator tubedown-100 tubedown-1; Gastric cancer antigen Ga19; N-alpha-acetyltransferase 15, NatA auxiliary subunit; N-terminal acetyltransferase; NMDA receptor regulated 1; NMDA receptor-regulated protein 1; Protein tubedown-1; Tbdn100; transcriptional coactivator tubedown-100; tubedown-1
Gene Aliases: GA19; NAA15; NARG1; NAT1P; NATH; TBDN; TBDN100
UniProt ID: (Human) Q9BXJ9
Entrez Gene ID: (Human) 80155
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.