Novus Biologicals
Manufacturer Code:NBP300272AF647
Catalog # NB087113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant NAA10 (P41227 Met1-Ser235). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARD1 ARD1 homolog A N-acetyltransferase (S. cerevisiae) ARD1 homolog N-acetyltransferase (S. cerevisiae) ARD1AARD1 homolog N-acetyltransferase EC 2.3.1.- EC 2.3.1.88 MGC71248 N(alpha)-acetyltransferase 10 NatA catalytic subunit N-acetyltransferase N-acetyltransferase ARD1 human homolog of N-alpha-acetyltransferase 10 NatA catalytic subunit N-terminal acetyltransferase complex ARD1 subunit homolog A; ARD1 homolog A, N-acetyltransferase; hARD1; N-acetyltransferase ARD1, human homolog of; N-alpha-acetyltransferase 10; N-alpha-acetyltransferase 10, NatA catalytic subunit; N-terminal acetyltransferase complex ARD1 subunit homolog A; NatA catalytic subunit Naa10
Gene Aliases: ARD1; ARD1A; ARD1P; DXS707; MCOPS1; NAA10; NATD; OGDNS; TE2
UniProt ID: (Human) P41227
Entrez Gene ID: (Human) 8260
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.