Invitrogen
Manufacturer Code:PA560017
Catalog # PIPA560017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human N6AMT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EEF1A lysine methyltransferase 1; N(6)-adenine-specific DNA methyltransferase 2; N-6 adenine-specific DNA methyltransferase 2 (putative); Protein-lysine N-methyltransferase N6AMT2; RP11-406H21.1 DMNT2 DNMT2 M.HsaIIP PuMet RNMT1
Gene Aliases: EEF1AKMT1; ESP13; N6AMT2
UniProt ID: (Human) Q8WVE0
Entrez Gene ID: (Human) 221143
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.