Invitrogen
Manufacturer Code:PA5110010
Catalog # PIPA5110010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-214 of human N6AMT2 (NP_777588.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2510005D08Rik AW045965 Ayu21-96 DMNT2 DNMT2 EEF1A lysine methyltransferase 1 EEF1AKMT1 ESP13 Gt(pU21)96Imeg GtAyu21-96 M.HsaIIP N(6)-adenine-specific DNA methyltransferase 2 N6amt2 Protein-lysine N-methyltransferase N6amt2 PuMet RGD1306762 RNMT1 RP11-406H21.1; EEF1A lysine methyltransferase 1; N(6)-adenine-specific DNA methyltransferase 2; N-6 adenine-specific DNA methyltransferase 2 (putative); Protein-lysine N-methyltransferase N6AMT2
Gene Aliases: EEF1AKMT1; ESP13; N6AMT2
UniProt ID: (Human) Q8WVE0
Entrez Gene ID: (Human) 221143
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.