Proteintech
Manufacturer Code:169541AP150UL
Catalog # 50-173-1291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | N6AMT2 Fusion Protein Ag10589 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EEF1A lysine methyltransferase 1; ESP13 N6AMT2; Gt(Ayu21)96Imeg; N(6)-adenine-specific DNA methyltransferase 2; N-6 adenine-specific DNA methyltransferase 2 (putative); Protein-lysine N-methyltransferase N6AMT2
Gene Aliases: 2510005D08Rik; AW045965; Ayu21-96; EEF1AKMT1; ESP13; Gt(pU21)96Imeg; GtAyu21-96; N6AMT2; RGD1306762
UniProt ID: (Human) Q8WVE0, (Mouse) Q9CY45
Entrez Gene ID: (Human) 221143, (Mouse) 68043, (Rat) 290279
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.